Isopentyl benzoate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.14500.0005 8.14500.0250
CA8.14500.0005EA 36 CAD
CA8.14500.0005 CA8.14500.0250
Isopentyl benzoate for synthesis, Sigma-Aldrich®
Isopentyl benzoate
Formula: C₁₂H₁₆O₂
MDL Number: MFCD00026515
CAS Number: 94-46-2

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Molar Mass 192.25 g/mol
Boiling Point 260 °C
Flash Point 89 °C
Refractive Index 1.4949 (20 °C, 589 nm)
Density 0.99 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR