(1S)-(-)-Borneol for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.14472.0025 8.14472.0100 8.14472.0250
CA8.14472.0025EA 179 CAD
CA8.14472.0025 CA8.14472.0100 CA8.14472.0250
(1S)-(-)-Borneol for synthesis, Sigma-Aldrich®
(1S)-(-)-Borneol
Formula: C₁₀H₁₈O
MDL Number: MFCD00003759
CAS Number: 464-45-9
UN: 1312
ADR: 4.1,III

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 205 - 208 °C
Molar Mass 154.25 g/mol
Boiling Point 210 °C (1036 hPa)
Flash Point 66 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR