Hydroxylamine 50% in water for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ200
8.14441.0100
8.14441.0500
CA8.14441.0100CS
723
CAD
CA8.14441.0100
CA8.14441.0500
Hydroxylamine 50% in water for synthesis, Sigma-Aldrich®
Hydroxylamine
Formula:
H₃NO Density: 1.12 g/cm3 (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00044522 CAS Number: 7803-49-8 EINECS: 232-259-2 UN: 3266 ADR: 8,III |
Specification Test Results
Assay (manganometric) | 47.0 - 53.0 % |
Density (d 20 °C/ 4 °C) | 1.120 - 1.123 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...