2-Ethylhexyl acetate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.14342.0250 8.14342.1000
CA8.14342.0250CS 548 CAD
CA8.14342.0250 CA8.14342.1000
2-Ethylhexyl acetate for synthesis, Sigma-Aldrich®
2-Ethylhexyl acetate
Formula: C₁₀H₂₀O₂
MDL Number: MFCD00027249
CAS Number: 103-09-3

Order Now

Specification Test Results

Solubility (20 °C) practically insoluble
Melting Point -92 °C
Molar Mass 172.26 g/mol
Boiling Point 199 °C (1013 hPa)
Vapor Pressure 0.28 hPa (20 °C)
Flash Point 85 °C
Refractive Index 1.4204 (20 °C, 589 nm)
Explosion Limit 0.76 - 8.14 %(V)
Density 0.872 g/cm3 (20 °C)
Ignition Point 268 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR