Saccharin sodium dihydrate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Sodium 3-oxo-3H-benzo[d]isothiazol-2-ide 1,1-dioxide dihydrate, Sodium Saccharin, Dihydrate, Saccharin sodium salt, o-sulphobenzimide sodium salt dihydrate, Saccharin Sodium Salt Dihydrate, o-Benzoic sulphimide sodium salt, o-sulfobenzimide sodium salt dihydrate, o-Benzoic sulfimide sodium salt

M-Clarity™ quality level = MQ200

8.14114.0100 8.14114.1000
CA8.14114.0100EA 53 CAD
CA8.14114.0100 CA8.14114.1000
Saccharin sodium dihydrate for synthesis, Sigma-Aldrich®
Saccharin sodium dihydrate
Formula: C₇H₅NNaO₃S·2H₂O
MW: 242.21 g/mol
MDL Number: MFCD00151213
CAS Number: 6155-57-3

Order Now

Specification Test Results

Solubility 1000 g/l (20 °C)
Molar Mass 241.19 g/mol
Bulk Density 600 - 800 kg/m3
Flash Point 200 °C
pH 6.0 - 7.5 (100 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR