2,6-Xylidine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: 2-Amino-m-xylene, 2,6-Dimethylaniline

M-Clarity™ quality level = MQ200

8.08671.0005 8.08671.0250 8.08671.1000
CA8.08671.0005EA 37 CAD
CA8.08671.0005 CA8.08671.0250 CA8.08671.1000
2,6-Xylidine for synthesis, Sigma-Aldrich®
2,6-Xylidine
Formula: C₈H₁₁N
MW: 121.18 g/mol
MDL Number: MFCD00007747
CAS Number: 87-62-7
UN: 1711
ADR: 6.1,II

Order Now

Specification Test Results

Solubility 13 g/l (20 °C)
Melting Point 10 - 12 °C
Molar Mass 121.18 g/mol
Boiling Point 216 °C (1013 hPa)
Vapor Pressure 0.2 hPa (20 °C)
Flash Point 92 °C
Refractive Index 1.5601 (20 °C, 589 nm)
Explosion Limit 1.3 - 6.9 %(V)
Density 0.98 g/cm3 (20 °C)
pH 12.5 (100 g/l, H2O, 20 °C)
Ignition Point 490 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR