Triphenylarsine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.08653.0005 8.08653.0025
CA8.08653.0005EA 74 CAD
CA8.08653.0005 CA8.08653.0025
Triphenylarsine for synthesis, Sigma-Aldrich®
Triphenylarsine
Formula: C₁₈H₁₅As
MDL Number: MFCD00002994
CAS Number: 603-32-7
UN: 3465
ADR: 6.1,II

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 59 - 61 °C
Molar Mass 306.24 g/mol
Boiling Point 360 °C (1013 hPa) (decomposition)
Flash Point 265 °C
Density 1.22 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR