Taurine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.08616.0005 8.08616.0250 8.08616.1000
CA8.08616.0005EA 47 CAD
CA8.08616.0005 CA8.08616.0250 CA8.08616.1000
Taurine for synthesis, Sigma-Aldrich®
Taurine
Formula: C₂H₇NO₃S
MDL Number: MFCD00008197
CAS Number: 107-35-7

Order Now

Specification Test Results

Solubility 65 g/l (12 °C)
Melting Point 300 °C (decomposition)
Molar Mass 125.15 g/mol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR