Titanyl(IV) acetylacetonate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Titanium(IV) oxide acetylacetonate, Titanium(IV) oxyacetylacetonate, Titanyl Acetylacetonate, Titanium (IV) oxide bis(acetylacetonate), Oxobis(2,4-pentanedionato-O,O')titanium (IV), Titanium (IV) oxide bis(2,4-pentanedionate)

M-Clarity™ quality level = MQ200

8.08305.0025 8.08305.0100
CA8.08305.0025EA 178 CAD
CA8.08305.0025 CA8.08305.0100
Titanyl(IV) acetylacetonate for synthesis, Sigma-Aldrich®
Titanyl (IV) acetylacetonate
Formula: C₁₀H₁₄O₅Ti
MW: 262.1 g/mol
MDL Number: MFCD00013505
CAS Number: 14024-64-7
EINECS: 237-861-9

Order Now

Specification Test Results

Solubility (20 °C) practically insoluble
Melting Point 200 °C
Molar Mass 262.08 g/mol
Bulk Density 500 kg/m3
Boiling Point 159 - 160 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR