1,2,3,4-Tetrahydronaphthalen-1-one (α-tetralone) for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: 3,4-Dihydronaphthalen-1(2H)-one

M-Clarity™ quality level = MQ200

8.08203.0100 8.08203.0250
CA8.08203.0100CS 1609 CAD
CA8.08203.0100 CA8.08203.0250
1,2,3,4-Tetrahydronaphthalen-1-one (α-tetralone) for synthesis, Sigma-Aldrich®
1,2,3,4-Tetrahydronaphthalen-1-one (1-Tetralone, α-Tetralone)
Formula: C₁₀H₁₀O
MW: 146.19 g/mol
MDL Number: MFCD00001688
CAS Number: 529-34-0
EINECS: 208-460-6

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 5 - 6 °C
Molar Mass 146.19 g/mol
Boiling Point 113 - 116 °C (8 hPa)
Flash Point 110 °C
Refractive Index 1.5685 (20 °C, 589 nm)
Density 1.10 g/cm3 (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR