Antimony(V) chloride for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.07837.0025 8.07837.0100
CA8.07837.0025EA 139 CAD
CA8.07837.0025 CA8.07837.0100
Antimony(V) chloride for synthesis, Sigma-Aldrich®
Antimony (V) chloride
Formula: Cl₅Sb
Boiling Pt: 140 °C (1013 hPa)
Melting Pt: 3 °C
Density: 2.35 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011213
CAS Number: 7647-18-9
EINECS: 231-601-8
UN: 1730
ADR: 8,II
Merck Index: 13,00701

Order Now

Specification Test Results

Solubility (20 °C) (rigorous decomposition)
Melting Point 3 - 4 °C
Molar Mass 299.01 g/mol
Boiling Point 140 °C
Vapor Pressure 1.1 hPa (20 °C)
Density 2.35 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR