Phenethylamine for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ200
8.07334.0005
8.07334.0250
CA8.07334.0005EA
47
CAD
CA8.07334.0005
CA8.07334.0250
Phenethylamine for synthesis, Sigma-Aldrich®
Phenethylamine
Formula:
C₆H₅CH₂CH₂NH₂ Boiling Pt: 200 °C (1013 hPa) Melting Pt: –60 °C Density: 0.9639 g/cm³ (20 °C) Flash Pt: 81 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008184 CAS Number: 64-04-0 EINECS: 200-574-4 UN: 2922 ADR: 8 (6.1),II Merck Index: 12,07371 |
Specification Test Results
Solubility | 4.3 g/l (20 °C) |
Molar Mass | 121.18 g/mol |
Boiling Point | 200 - 202 °C (1013 hPa) |
Vapor Pressure | 0.4 hPa (20 °C) |
Flash Point | 81 °C |
Refractive Index | 1.5332 (20 °C, 589 nm) |
Explosion Limit | 1.0 - 5.5 %(V) |
Density | 0.96 g/cm3 (20 °C) |
pH | 11.5 (4.3 g/l, H2O, 20 °C) |
Ignition Point | 425 °C |
Viscosity kinematic | 2.40 mm2/s (23 °C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...