Pelargonic acid for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
M-Clarity™ quality level = MQ200
8.07167.0100
8.07167.1000
CA8.07167.0100CS
773
CAD
CA8.07167.0100
CA8.07167.1000
Pelargonic acid for synthesis, Sigma-Aldrich®
Pelargonic acid
Formula:
C₉H₁₈O₂ Boiling Pt: 253 °C (1013 hPa) Melting Pt: 12 °C Density: 0.9055 g/cm³ (20 °C) Flash Pt: 133 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00004433 CAS Number: 112-05-0 EINECS: 203-931-2 UN: 3265 ADR: 8,I Merck Index: 12,07198 |
Specification Test Results
Solubility | 0.3 g/l (25 °C) |
Melting Point | 12 °C |
Molar Mass | 158.24 g/mol |
Boiling Point | 252 - 255 °C (1013 hPa) |
Vapor Pressure | 0.04 hPa (20 °C) |
Flash Point | 133 °C |
Refractive Index | 1.4330 (20 °C, 589 nm) |
Explosion Limit | 0.8 - 9 %(V) |
Density | 0.91 g/cm3 (20 °C) |
pH |
3.5 ( |
Ignition Point | 405 °C |
Viscosity kinematic | 9.0 mm2/s (20 °C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...