2-Nitrotoluene for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.06806.0100 8.06806.1000
CA8.06806.0100CS 742 CAD
CA8.06806.0100 CA8.06806.1000
2-Nitrotoluene for synthesis, Sigma-Aldrich®
2-Nitrotoluene
Formula: C₇H₇NO₂
MW: 137.14 g/mol
Boiling Pt: 221.7
Melting Pt: -1
Density: 1.163 g/cm³ (20 °C)
Flash Pt: 106
Storage Temperature: Ambient
MDL Number: MFCD00007157
CAS Number: 88-72-2
EINECS: 201-853-3
UN: 1664
ADR: 6.1 II
Merck Index: 12,06748

Order Now

Specification Test Results

Solubility 0.65 g/l (20 °C) (experimental)
Melting Point -4 - -3 °C
Molar Mass 137.13 g/mol
Boiling Point 221 - 222 °C (1013 hPa)
Vapor Pressure 0.1 hPa (20 °C)
Flash Point 106 °C
Refractive Index 1.545 (20 °C, 589 nm)
Explosion Limit 1.47 - 8.8 %(V)
Density 1.16 g/cm3 (20 °C)
pH 6 - 8 (H2O)
Ignition Point 420 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR