4-Nitrobenzaldehyde for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.06766.0025 8.06766.0100
CA8.06766.0025EA 77 CAD
CA8.06766.0025 CA8.06766.0100
4-Nitrobenzaldehyde for synthesis, Sigma-Aldrich®
4-Nitrobenzaldehyde
Formula: C₇H₅NO₃
Boiling Pt: 300 °C (1013 hPa)
Melting Pt: 106 °C
Density: 1.546 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007346
CAS Number: 555-16-8
EINECS: 209-084-5
Merck Index: 12,06684

Order Now

Specification Test Results

Solubility 2.34 g/l (25 °C)
Melting Point 101 - 104 °C
Molar Mass 151.12 g/mol
Bulk Density 400 kg/m3


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR