2-Pentanone for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ200
8.05965.0100
8.05965.0500
CA8.05965.0100CS
1006
CAD
CA8.05965.0100
CA8.05965.0500
2-Pentanone for synthesis, Sigma-Aldrich®
2-Pentanone
Formula:
C₅H₁₀O Boiling Pt: 100 °C (1013 hPa) Melting Pt: –78 °C Density: 0.809 g/cm³ (20 °C) Flash Pt: 7 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009400 CAS Number: 107-87-9 EINECS: 203-528-1 UN: 1249 ADR: 3,II Merck Index: 12,06193 |
Specification Test Results
Solubility | 43 g/l (25 °C) |
Melting Point | -78 °C |
Molar Mass | 86.13 g/mol |
Boiling Point | 101 - 103 °C (1013 hPa) |
Vapor Pressure | 37 hPa (20 °C) |
Flash Point | 7 °C |
Refractive Index | 1.3895 (20 °C, 589 nm) |
Explosion Limit | 1.5 - 8.2 %(V) |
Density | 0.81 g/cm3 (20 °C) |
Ignition Point | 449 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...