(±)-4-Methyl-2-pentanol for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
M-Clarity™ quality level = MQ200
8.05804.0100
8.05804.1000
CA8.05804.0100CS
2108
CAD
CA8.05804.0100
CA8.05804.1000
(±)-4-Methyl-2-pentanol for synthesis, Sigma-Aldrich®
(±)-4-Methyl-2-pentanol
Formula:
C₆H₁₄O Boiling Pt: 132 °C (1013 hPa) Melting Pt: –90 °C Density: 0.8093 g/cm³ (20 °C) Flash Pt: 54 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004550 CAS Number: 108-11-2 EINECS: 203-551-7 UN: 2053 ADR: 3,III |
Specification Test Results
Solubility | 16.4 g/l (20 °C) |
Melting Point | -60 °C |
Molar Mass | 102.18 g/mol |
Boiling Point | 130 - 133 °C (1013 hPa) |
Vapor Pressure | 7.06 hPa (25 °C) |
Flash Point | 41 °C |
Explosion Limit | 1 - 5.5 %(V) |
Density | 0.81 g/cm3 (20 °C) |
Ignition Point | 305 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...