1-Iodopropane stabilized for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: 1-Propyl iodide

Stabilization: Stabilised with sodium carbonate (Na₂CO₃)
M-Clarity™ quality level = MQ200

8.04788.0100 8.04788.0250
CA8.04788.0100EA 144 CAD
CA8.04788.0100 CA8.04788.0250
1-Iodopropane stabilized for synthesis, Sigma-Aldrich®
1-Iodopropane
Formula: C₃H₇I
MW: 169.99 g/mol
Boiling Pt: 103 °C (1013 hPa)
Melting Pt: –98 °C
Density: 1.741 g/cm³ (20 °C)
Flash Pt: 44 °C
Storage Temperature: Ambient
MDL Number: MFCD00001095
CAS Number: 107-08-4
EINECS: 203-460-2
UN: 2392
ADR: 3,III
Merck Index: 12,08047

Order Now

Specification Test Results

Solubility 1.1 g/l (20 °C)
Melting Point -98 °C
Molar Mass 169.99 g/mol
Boiling Point 103 °C (1013 mbar)
Vapor Pressure 57 hPa (25 °C)
Flash Point 44 °C
Refractive Index 1.5055 (20 °C, 589 nm)
Density 1.75 g/cm3 (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR