1-Iodobutane for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ200
8.04786.0100
8.04786.0250
CA8.04786.0100CS
1693
CAD
CA8.04786.0100
CA8.04786.0250
1-Iodobutane for synthesis, Sigma-Aldrich®
1-Iodobutane
Formula:
C₄H₉I Boiling Pt: 130 °C (1013 hPa) Melting Pt: –103 °C Density: 1.616 g/cm³ (20 °C) Flash Pt: 36 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001098 CAS Number: 542-69-8 EINECS: 208-824-4 UN: 1993 ADR: 3,II Merck Index: 13,01572 |
Specification Test Results
Solubility | 0.2 g/l (20 °C) |
Melting Point | -103 °C |
Molar Mass | 184.02 g/mol |
Boiling Point | 130 °C (1013 hPa) |
Vapor Pressure | 18.5 hPa (25 °C) |
Flash Point | 33 °C |
Refractive Index | 1.4979 (20 °C, 589 nm) |
Density | 1.62 g/cm3 (20 °C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...