2,4-Dichlorophenol for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.03774.0100 8.03774.0500
CA8.03774.0100CS 1292 CAD
CA8.03774.0100 CA8.03774.0500
2,4-Dichlorophenol for synthesis, Sigma-Aldrich®
2,4-Dichlorophenol
Formula: C₆H₄Cl₂O
Boiling Pt: 210 °C (1013 hPa)
Melting Pt: 40…41 °C
Density: 1.4 g/cm³ (20 °C)
Flash Pt: 114 °C
Storage Temperature: Ambient
MDL Number: MFCD00002169
CAS Number: 120-83-2
EINECS: 204-429-6
UN: 2020
ADR: 6.1,III
Merck Index: 13,03098

Order Now

Specification Test Results

Solubility 4.5 g/l (20 °C)
Melting Point 40 - 43 °C
Molar Mass 163 g/mol
Boiling Point 209 - 211 °C (1013 hPa)
Vapor Pressure 1.3 hPa (50 °C)
Flash Point 114 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR