Dichloromethyl methyl ether for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ200
8.03467.0025
8.03467.0100
CA8.03467.0025EA
252
CAD
CA8.03467.0025
CA8.03467.0100
Dichloromethyl methyl ether for synthesis, Sigma-Aldrich®
Dichloromethyl methyl ether
Formula:
C₂H₄Cl₂O Density: 1.29 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00000828 CAS Number: 4885-02-3 EINECS: 225-498-9 UN: 2924 ADR: 3 (8),II |
Specification Test Results
Solubility | (20 °C) (decomposition) |
Molar Mass | 114.95 g/mol |
Boiling Point | 85 - 87 °C (1013 hPa) |
Vapor Pressure | 84 hPa (20 °C) |
Flash Point | 42 °C |
Refractive Index | 1.43 (20 °C, 589 nm) |
Density | 1.29 g/cm3 (20 °C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...