2,4-Diaminotoluene for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.03269.0005 8.03269.0250 8.03269.1000
CA8.03269.0005EA 45 CAD
CA8.03269.0005 CA8.03269.0250 CA8.03269.1000
2,4-Diaminotoluene for synthesis, Sigma-Aldrich®
2,4-Diaminotoluene
Formula: C₇H₁₀N₂
Boiling Pt: 292 °C (1013 hPa)
Melting Pt: 99 °C
Density: 1.045 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007804
CAS Number: 95-80-7
EINECS: 202-453-1
UN: 1709
ADR: 6.1,III

Order Now

Specification Test Results

Solubility 35 g/l (20 °C)
Melting Point 96 - 99 °C
Molar Mass 122.17 g/mol
Bulk Density 500 kg/m3
Boiling Point 284 - 286 °C (1013 hPa)
Vapor Pressure 1 hPa (106 °C)
Flash Point 169 °C
Ignition Point 520 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR