N,N-Diethylethylenediamine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.03215.0005 8.03215.0250
CA8.03215.0005EA 37 CAD
CA8.03215.0005 CA8.03215.0250
N,N-Diethylethylenediamine for synthesis, Sigma-Aldrich®
N,N-Diethylethylenediamine
Formula: C₆H₁₆N₂
Boiling Pt: 146 °C (1013 hPa)
Melting Pt: –70 °C
Density: 0.82 g/cm³ (20 °C)
Flash Pt: 36 °C
MDL Number: MFCD00008176
CAS Number: 100-36-7
EINECS: 202-844-7
UN: 2685
ADR: 8 (3),II

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -70 °C
Molar Mass 116.21 g/mol
Boiling Point 145 - 148 °C (1013 hPa)
Flash Point 36 °C
Explosion Limit 0.7 - 8.4 %(V)
Density 0.82 g/cm3 (20 °C)
Ignition Point 245 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR