N,N-Diethylacetamide for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.03214.0025 8.03214.0100
CA8.03214.0025EA 87 CAD
CA8.03214.0025 CA8.03214.0100
N,N-Diethylacetamide for synthesis, Sigma-Aldrich®
N,N-Diethylacetamide
Formula: C₆H₁₃NO
Boiling Pt: 182 °C (1013 hPa)
Density: 1.02 g/cm³ (20 °C)
Flash Pt: 76 °C
Storage Temperature: Ambient
MDL Number: MFCD00009047
CAS Number: 685-91-6
EINECS: 211-685-2

Order Now

Specification Test Results

Solubility 53.5 g/l (25 °C)
Molar Mass 115.17 g/mol
Boiling Point 182 - 186 °C (1013 hPa)
Vapor Pressure 1.2 hPa (25 °C)
Flash Point 76 °C
Refractive Index 1.4401 (20 °C, 589 nm)
Density 0.91 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR