Dicyclohexylamine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: DCHA

M-Clarity™ quality level = MQ200

8.02948.0100 8.02948.1000
CA8.02948.0100CS 793 CAD
CA8.02948.0100 CA8.02948.1000
Dicyclohexylamine for synthesis, Sigma-Aldrich®
Dicyclohexylamine
Formula: C₁₂H₂₃N
MW: 181.32 g/mol
Boiling Pt: 255.8 °C (1013 hPa)
Melting Pt: –2 °C
Density: 0.91 g/cm³ (20 °C)
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00011658
CAS Number: 101-83-7
EINECS: 202-980-7
UN: 2565
ADR: 8,III
Merck Index: 13,03122

Order Now

Specification Test Results

Solubility 1 g/l (20 °C)
Melting Point -0.1 °C
Molar Mass 181.32 g/mol
Boiling Point 256 °C (1013 hPa)
Vapor Pressure 0.04 hPa (25 °C)
Flash Point 105 °C
Refractive Index 1.4842 (20 °C, 589 nm)
Explosion Limit 0.8 - 4.6 %(V)
Density 0.91 g/cm3 (20 °C)
pH 11 (1 g/l, H2O, 20 °C)
Ignition Point 240 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR