Acetonedimethyl acetal for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: 2,2-Dimethoxypropane, Dimethoxypropane, DMP, Dimethoxy-2,2-propane

M-Clarity™ quality level = MQ200

8.02936.0100 8.02936.0500 8.02936.1000
CA8.02936.0100EA 49 CAD
CA8.02936.0100 CA8.02936.0500 CA8.02936.1000
Acetonedimethyl acetal for synthesis, Sigma-Aldrich®
Acetone-dimethyl acetal
Formula: C₅H₁₂O₂
Boiling Pt: 83 °C (1013 hPa)
Melting Pt: –47 °C
Density: 0.845 g/cm³ (20 °C)
Flash Pt: –5 °C
Storage Temperature: Ambient
MDL Number: MFCD00008479
CAS Number: 77-76-9
EINECS: 201-056-0
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Solubility 180 g/l (20 °C)
Melting Point -47 °C
Molar Mass 104.15 g/mol
Boiling Point 80 °C
Vapor Pressure 60 hPa (16 °C)
Flash Point -10 °C
Refractive Index 1.378 (20 °C, 589 nm)
Explosion Limit 6 - 31 %(V)
Density 0.85 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR