Bromoacetic acid ≥98.0% (by GC) for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.02260.0005 8.02260.0250 8.02260.1000
CA8.02260.0005EA 40 CAD
CA8.02260.0005 CA8.02260.0250 CA8.02260.1000
Bromoacetic acid ≥98.0% (by GC) for synthesis, Sigma-Aldrich®
Bromoacetic acid
Formula: BrCH₂COOH
Boiling Pt: 206 °C (1013 hPa)
Melting Pt: 46…49 °C
Density: 1.9335 g/cm³ (20 °C)
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00002678
CAS Number: 79-08-3
EINECS: 201-175-8
UN: 3425
ADR: 8,II
Merck Index: 13,01384

Order Now

Specification Test Results

Solubility 93.8 g/l (25 °C)
Melting Point 46 - 49 °C
Molar Mass 138.94 g/mol
Boiling Point 208 °C (1013 hPa)
Vapor Pressure 0.158 hPa (25 °C)
Flash Point 112 °C
Density 1.93 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR