Ethyl decanoate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Ethyl caprate, Capric acid ethyl ester, Decanoic acid ethyl ester, Decylic acid ethyl ester

M-Clarity™ quality level = MQ200

8.02180.0005 8.02180.0250
CA8.02180.0005EA 40 CAD
CA8.02180.0005 CA8.02180.0250
Ethyl decanoate for synthesis, Sigma-Aldrich®
Ethyl decanoate
Formula: C₁₂H₂₄O₂
MW: 200.32 g/mol
Boiling Pt: 245
Melting Pt: –20 °C
Density: 0.862 g/cm³ (20 °C)
MDL Number: MFCD00009581
CAS Number: 110-38-3
EINECS: 203-761-9
Merck Index: 13,03811

Order Now

Specification Test Results

Solubility 0.016 g/l (20 °C)
Melting Point -20 °C
Molar Mass 200.32 g/mol
Boiling Point 244 - 246 °C (1013 hPa)
Vapor Pressure 0.02 hPa (20 °C)
Flash Point 104 °C
Refractive Index 1.4248 (20 °C, 589 nm)
Explosion Limit 0.7 %(V)
Density 0.86 g/cm3 (20 °C)
Ignition Point 310 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR