Butyryl chloride for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.01977.0250 8.01977.1000
CA8.01977.0250CS 906 CAD
CA8.01977.0250 CA8.01977.1000
Butyryl chloride for synthesis, Sigma-Aldrich®
Butyryl chloride
Formula: C₄H₇ClO
Boiling Pt: 96…102 °C (1013 hPa)
Melting Pt: –89 °C
Density: 1.0277 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000752
CAS Number: 141-75-3
EINECS: 205-498-5
UN: 2353
ADR: 3 (8),II
Merck Index: 13,01598

Order Now

Specification Test Results

Solubility (20 °C) decomposes,Hydrolysis
Melting Point -89 °C
Molar Mass 106.55 g/mol
Boiling Point 101 - 105 °C (1013 hPa)
Vapor Pressure 39 hPa (20 °C)
Flash Point 18 °C
Refractive Index 1.412 (20 °C, 589 nm)
Explosion Limit 2.5 - 7.3 %(V)
Density 1.03 g/cm3 (20 °C)
pH (H2O) acidic
Ignition Point 280 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR