Benzyl chloride for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: (Chloromethyl)benzene, Chlorophenylmethane, α-Chlorotoluene

M-Clarity™ quality level = MQ200

8.01809.0100 8.01809.1000 8.01809.2500
CA8.01809.0100CS 652 CAD
CA8.01809.0100 CA8.01809.1000 CA8.01809.2500
Benzyl chloride for synthesis, Sigma-Aldrich®
Benzyl chloride
Formula: (C₆H₅)CH₂Cl
Boiling Pt: 179.3 °C (1013 hPa)
Melting Pt: –41.2 °C
Density: 1.103 g/cm³ (20 °C)
Flash Pt: 52 °C
Storage Temperature: Ambient
MDL Number: MFCD00000889
CAS Number: 100-44-7
EINECS: 202-853-6
UN: 1738
ADR: 6.1 (8),II
Merck Index: 13,01131

Order Now

Specification Test Results

Solubility 460 mg/l (30 °C)
Melting Point -41.2 °C
Molar Mass 126.58 g/mol
Boiling Point 179.3 °C (1013 hPa)
Vapor Pressure 1.2 hPa (20 °C)
Flash Point 67 °C
Refractive Index 1.5415
Explosion Limit 1.1 - 14 %(V)
Density 1.10 g/cm3 (20 °C)
pH (H2O) not applicable
Ignition Point 585 °C
Hazard Class 6.1
Hazard Class Description Toxic materials


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR