Methyl formate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.00889.0100 8.00889.1000 8.00889.2500
CA8.00889.0100CS 700 CAD
CA8.00889.0100 CA8.00889.1000 CA8.00889.2500
Methyl formate for synthesis, Sigma-Aldrich®
Methyl formate
Formula: C₂H₄O₂
Boiling Pt: 32 °C (1013 hPa)
Melting Pt: –100.4 °C
Density: 0.9713 g/cm³ (20 °C)
Flash Pt: –32 °C
Storage Temperature: Ambient
MDL Number: MFCD00003291
CAS Number: 107-31-3
EINECS: 203-481-7
UN: 1243
ADR: 3,I
Merck Index: 12,06153

Order Now

Specification Test Results

Solubility 300 g/l (20 °C)
Melting Point -100.4 °C
Molar Mass 60.05 g/mol
Boiling Point 31.5 °C (1013 hPa)
Vapor Pressure 644 hPa (20 °C)
Flash Point -32 °C
Refractive Index 1.3433 (20 °C, 589 nm)
Explosion Limit 5 - 23 %(V)
Density 0.968 g/cm3 (20 °C)
pH 4 - 5 (200 g/l, H2O, 20 °C)
Ignition Point 450 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR