2-Ethylhexyl acrylate stabilized for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


Stabilization: Stabilised with hydroquinone monomethyl ether (4-methoxyphenol, MEHQ)
M-Clarity™ quality level = MQ200

8.00835.0100 8.00835.0500
CA8.00835.0100CS 563 CAD
CA8.00835.0100 CA8.00835.0500
2-Ethylhexyl acrylate stabilized for synthesis, Sigma-Aldrich®
2-Ethylhexyl acrylate
Formula: C₁₁H₂₀O₂
MDL Number: MFCD00009495
CAS Number: 103-11-7
EINECS: 203-080-7

Order Now

Specification Test Results

Solubility 0.1 g/l (25 °C)
Melting Point -90 °C
Molar Mass 184.27 g/mol
Boiling Point 229 °C (1013 hPa)
Vapor Pressure 0.12 hPa (20 °C)
Flash Point 86 °C
Explosion Limit 0.9 - 6.0 %(V)
Density 0.887 g/cm3 (20 °C)
Ignition Point 230 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR