Butyl acrylate stabilized for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


Stabilization: Stabilised with hydroquinone monomethyl ether (4-methoxyphenol, MEHQ)
M-Clarity™ quality level = MQ200

8.00832.0100 8.00832.0500 8.00832.2500
CA8.00832.0100CS 437 CAD
CA8.00832.0100 CA8.00832.0500 CA8.00832.2500
Butyl acrylate stabilized for synthesis, Sigma-Aldrich®
Butyl acrylate
Formula: C₇H₁₂O₂
MDL Number: MFCD00008809
CAS Number: 141-32-2
EINECS: 205-480-7
UN: 2348
ADR: 3,III

Order Now

Specification Test Results

Solubility 2 g/l (20 °C)
Melting Point -65 °C
Molar Mass 128.17 g/mol
Boiling Point 147 - 148 °C (1013 hPa)
Vapor Pressure 4.3 hPa (20 °C)
Flash Point 36.5 °C
Refractive Index 1.419 (20 °C, 589 nm)
Explosion Limit 1.1 - 7.8 %(V)
Density 0.90 g/cm3 (20 °C)
Ignition Point 267 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR