Sebacic acid for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Decanedioic acid, 1,8-Octanedicarboxylic acid, 1,10-Decanedioic acid

M-Clarity™ quality level = MQ200

8.00753.0100 8.00753.0500 8.00753.1000
CA8.00753.0100CS 625 CAD
CA8.00753.0100 CA8.00753.0500 CA8.00753.1000
Sebacic acid for synthesis, Sigma-Aldrich®
Sebacic acid
Formula: C₁₀H₁₈O₄
MW: 202.25 g/mol
Boiling Pt: 374
Melting Pt: 127…131 °C
Density: 1.21 g/cm³ (20 °C)
Flash Pt: 221
Storage Temperature: Ambient
MDL Number: MFCD00004440
CAS Number: 111-20-6
EINECS: 203-845-5
REACH: 01-2119519212-52
Merck Index: 12,08558

Order Now

Specification Test Results

Solubility 1 g/l (20 °C)
Melting Point 130 - 133 °C
Molar Mass 202.24 g/mol
Bulk Density 600 - 620 kg/m3
Boiling Point 295 °C (133 hPa)
Vapor Pressure 1.3 hPa (183 °C)
Density 1.21 g/cm3


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR