3,5-Dihydroxybenzoic acid for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: alpha-Resorcylic acid

M-Clarity™ quality level = MQ200

8.00737.0100 8.00737.0250
CA8.00737.0100CS 1191 CAD
CA8.00737.0100 CA8.00737.0250
3,5-Dihydroxybenzoic acid for synthesis, Sigma-Aldrich®
3,5-Dihydroxybenzoic acid
Formula: C₇H₆O₄
MW: 154.12 g/mol
Melting Pt: 236…238 °C
Flash Pt: >200 °C
Storage Temperature: Ambient
MDL Number: MFCD00002512
CAS Number: 99-10-5
EINECS: 202-730-7

Order Now

Specification Test Results

Solubility 84 g/l (20 °C)
Melting Point 236 - 238 °C
Molar Mass 154.12 g/mol
Bulk Density 700 kg/m3
Density 2.3 (10 g/l, H2O, 25 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR