Butyl methacrylate stabilised for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: Methacrylic acid butyl ester

M-Clarity™ quality level = MQ200

8.00585.0100 8.00585.0500
CA8.00585.0100CS 1038 CAD
CA8.00585.0100 CA8.00585.0500
Butyl methacrylate stabilised for synthesis, Sigma-Aldrich®
Butyl methacrylate
Formula: C₈H₁₄O₂
MW: 142.2 g/mol
Boiling Pt: 163
Melting Pt: -75
Density: 0.8951 g/cm³ (20 °C)
MDL Number: MFCD00009444
CAS Number: 97-88-1
EINECS: 202-615-1
UN: 2227
ADR: 3 III

Order Now

Specification Test Results

Solubility 0.36 g/l (25 °C)
Melting Point -75 °C
Molar Mass 142.20 g/mol
Boiling Point 163 °C (1013 hPa)
Vapor Pressure 2.7 hPa (20 °C)
Flash Point 46 °C
Refractive Index 1.424 (20 °C, 589 nm)
Explosion Limit 2.00 - 8.00 %(V)
Density 0.89 g/cm3 (20 °C)
Ignition Point 290 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR