p-Anisidine for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
4-Anisidine, 4-Aminoanisole, 4-Methoxyaniline
M-Clarity™ quality level = MQ200
8.00458.0100
8.00458.0500
CA8.00458.0100EA
60
CAD
CA8.00458.0100
CA8.00458.0500
p-Anisidine for synthesis, Sigma-Aldrich®
p-Anisidine
Formula:
C₇H₉NO Boiling Pt: 243 °C (1013 hPa) Melting Pt: 55…58 °C Density: 1.2 g/cm³ (20 °C) Flash Pt: 122 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007864 CAS Number: 104-94-9 EINECS: 203-254-2 UN: 3143 ADR: 6.1,III Merck Index: 13,00670 |
Specification Test Results
Solubility | 21 g/l (20 °C) |
Melting Point | 55 - 58 °C |
Molar Mass | 123.15 g/mol |
Bulk Density | 700 kg/m3 |
Boiling Point | 243 °C |
Vapor Pressure | 0.02 hPa (20 °C) |
Flash Point | 122 °C |
Density | 1.06 g/cm3 (63 °C) |
pH | 7.7 (1 g/l, H2O, 20 °C) |
Ignition Point | 450 °C |
Hazard Class | 6.1 |
Hazard Class Description | Toxic materials |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...