(±)-Allyl-2,3-epoxypropyl ether for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Allyl glycidyl ether

M-Clarity™ quality level = MQ200

8.00259.0250 8.00259.2500
CA8.00259.0250CS 1778 CAD
CA8.00259.0250 CA8.00259.2500
(±)-Allyl-2,3-epoxypropyl ether for synthesis, Sigma-Aldrich®
(±)-Allyl 2,3-epoxypropyl ether
Formula: C₆H₁₀O₂
MW: 114.14 g/mol
MDL Number: MFCD00005143
CAS Number: 106-92-3
EINECS: 203-442-4
UN: 2219
ADR: 3 III

Order Now

Specification Test Results

Solubility 50 g/l (20 °C)
Melting Point -100 °C
Molar Mass 114.15 g/mol
Boiling Point 154 °C (1013 hPa)
Vapor Pressure 2.6 hPa (20 °C)
Flash Point 45 °C
Refractive Index 1.4332 (20 °C, 589 nm)
Density 0.97 g/cm3 (20 °C)
Ignition Point 264 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR