4-Hydroxycinnamic acid ≥98% for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: p-Coumaric acid, p-Cumaric acid, 3-(4-Hydroxyphenyl)acrylic acid, p-Hydroxy-Cinnamic Acid

M-Clarity™ quality level = MQ200

8.00237.0010 8.00237.0050
CA8.00237.0010EA 75 CAD
CA8.00237.0010 CA8.00237.0050
4-Hydroxycinnamic acid ≥98% for synthesis, Sigma-Aldrich®
4-Hydroxycinnamic acid
Formula: C₉H₈O₃
MDL Number: MFCD00004399
CAS Number: 7400-08-0
EINECS: 231-000-0

Order Now

Specification Test Results

Solubility (20 °C) slightly soluble
Melting Point 215 - 220 °C
Molar Mass 164.16 g/mol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR