Caproic acid for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.00198.0005 8.00198.0250 8.00198.1000
CA8.00198.0005EA 40 CAD
CA8.00198.0005 CA8.00198.0250 CA8.00198.1000
Caproic acid for synthesis, Sigma-Aldrich®
Caproic acid
Formula: C₆H₁₂O₂
Boiling Pt: 202 °C (1013 hPa)
Melting Pt: –4.8 °C
Density: 0.9276 g/cm³ (20 °C)
Flash Pt: 102 °C
Storage Temperature: Ambient
MDL Number: MFCD00004421
CAS Number: 142-62-1
EINECS: 205-550-7
UN: 2829
ADR: 8,III
Merck Index: 13,01765

Order Now

Specification Test Results

Solubility 9.7 g/l (20 °C)
Melting Point -3 °C
Molar Mass 116.16 g/mol
Boiling Point 206 °C (1013 hPa)
Vapor Pressure 0.3 hPa (20 °C)
Flash Point 110 °C
Refractive Index 1.4162 (20 °C, 589 nm)
Density 0.93 g/cm3 (20 °C)
pH 4 (1 g/l, H2O, 20 °C)
Ignition Point 380 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR