Diethyl adipate for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
Synonyms:
NSC 19160, Adipic acid diethyl ester, 1,6-Hexanedioic acid diethyl ester, NSC 3363
M-Clarity™ quality level = MQ200
8.00086.0100
8.00086.0500
CA8.00086.0100CS
773
CAD
CA8.00086.0100
CA8.00086.0500
Diethyl adipate for synthesis, Sigma-Aldrich®
Diethyl adipate
Formula:
C₁₀H₁₈O₄ Boiling Pt: ∼251 °C (1013 hPa) Melting Pt: –19.8 °C Density: 1.01 g/cm³ (20 °C) Flash Pt: >100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009215 CAS Number: 141-28-6 EINECS: 205-477-0 Merck Index: 13,00163 |
Specification Test Results
Solubility |
|
Melting Point | -19.8 °C |
Molar Mass | 202.25 g/mol |
Boiling Point | 245 °C (1013 hPa) |
Flash Point |
|
Density | 1.01 g/cm3 (20 °C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...