Water, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma

Synonyms: Hydrogen oxide

M-Clarity™ quality level = MQ300

1.16754.4000 1.16754.5000 1.16754.9010
CA1.16754.4000CS 871 CAD
CA1.16754.4000 CA1.16754.5000 CA1.16754.9010
Water, EMSURE® for analysis, Supelco®
Water
Formula: H₂O
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5

Order Now

Specification Test Results

Molar Mass 18.02 g/mol
Boiling Point 100 °C (1013 hPa)
Vapor Pressure 23 hPa (20 °C)
Density 1.00 g/cm3 (20 °C)
pH (H2O, 20 °C) neutral


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR