Tetrahydrofuran ≥99.8% (by GC), EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Danger

1.09731.1000 1.09731.2500 1.09731.6010 1.09731.6025
CA1.09731.1000CS 1373 CAD
CA1.09731.1000 CA1.09731.2500 CA1.09731.6010 CA1.09731.6025
Tetrahydrofuran ≥99.8% (by GC), EMSURE® for analysis, Supelco®
Tetrahydrofuran
Formula: C₄H₈O
Boiling Pt: 65…67 °C (lit.)
Melting Pt: –108 °C (lit.)
Density: 0.889 g/ml (25°C) (lit.)
Flash Pt: –21.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00005356
CAS Number: 109-99-9
EINECS: 203-726-8
UN: 2056
ADR: 3,II
REACH: 05-2114615188-48
Merck Index: 12,09351
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2d7611caf0127fbef941994edd1d7e69487d54a&vid=2a64e21bd2e13636c3038adf839b3e9ff18509c5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -108.5 °C
Molar Mass 72.11 g/mol
Boiling Point 65 - 66 °C
Vapor Pressure 173 hPa (20 °C)
Flash Point -21.5 °C
Refractive Index 1.407 (20 °C)
Explosion Limit 1.5 - 12.4 %(V)
Density 0.89 g/cm3 (20 °C)
pH 7 - 8 (200 g/l, H2O, 20 °C)
Ignition Point 215 °C
Evaporation number 2.3


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR