Petroleum, EMSURE® analysis_for, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Kerosene

1.09718.1000 1.09718.2500 1.09718.6025
CA1.09718.1000CS 1022 CAD
CA1.09718.1000 CA1.09718.2500 CA1.09718.6025
Petroleum, EMSURE® analysis_for, Supelco®
Petroleum
Melting Pt: <–50 °C
Density: 0.749…0.761 g/cm3 (25 °C)
Storage Temperature: Ambient
CAS Number: 64742-48-9
UN: 3295
ADR: 3 III

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Boiling Point 172 - 215 °C (1013 hPa)
Vapor Pressure 1.0 hPa (20 °C)
Flash Point 56 °C
Explosion Limit 0.6 - 8 %(V)
Density 0.76 g/cm3 (20 °C)
Ignition Point 355 °C
Viscosity kinematic 1.85 mm2/s (25 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR