Formamide, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®

Supplier: MilliporeSigma
Danger

1.09684.1000 1.09684.2500
CA1.09684.1000CS 4039 CAD
CA1.09684.1000 CA1.09684.2500
Formamide, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Formamide
Formula: HCONH₂
Boiling Pt: 210 °C (1013 hPa)
Melting Pt: 2 °C
Density: 1.1334 g/cm³ (20 °C)
Flash Pt: 79 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007941
CAS Number: 75-12-7
EINECS: 200-842-0
Merck Index: 13,04261
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f381691a96baf5054ca8291e1fa9e9603b687e31&vid=e54dda1a985d7a4f9485c2cc5233498087e43f88&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 2.9 °C
Molar Mass 45.04 g/mol
Boiling Point 210 °C (1013 hPa)
Vapor Pressure 0.08 hPa (20 °C)
Flash Point 175 °C
Explosion Limit 2.7 - 19.0 %(V)
Density 1.13 g/cm3 (20 °C)
pH 8 - 10 (200 g/l, H2O, 20 °C)
Ignition Point 500 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR