1,4-Dioxane ≥99.5% (by GC), EMSURE® ACS, ISO for analysis, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Glycolethylether, 1,4-Dioxacyclohexane, Dioxane, p-Dioxane, 1,4-Diethylene dioxide

1.09671.0250 1.09671.1000 1.09671.2500 1.09671.6025
CA1.09671.0250CS 1294 CAD
CA1.09671.0250 CA1.09671.1000 CA1.09671.2500 CA1.09671.6025
1,4-Dioxane ≥99.5% (by GC), EMSURE® ACS, ISO for analysis, Supelco®
1,4-Dioxane
Formula: C₄H₈O₂
MW: 88.11 g/mol
Boiling Pt: 101.5 °C (1013 hPa)
Melting Pt: 12 °C
Density: 1.03 g/cm3 (20 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00006571
CAS Number: 123-91-1
EINECS: 204-661-8
UN: 1165
ADR: 3,II
Merck Index: 13,03330
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df424642d4a7a96d2eebfb99cf9a2e6d128185d0&vid=37c3b8119f755f99624576b4e975000cbbb8349d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 12 °C
Molar Mass 88.11 g/mol
Boiling Point 101.5 °C (1013 hPa)
Vapor Pressure 41 hPa (20 °C)
Flash Point 11 °C
Explosion Limit 1.7 - 25.2 %(V)
Density 1.03 g/cm3 (20 °C)
pH 6 - 8 (500 g/l, H2O, 20 °C)
Ignition Point 300 °C
Evaporation number 7.3

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR