Cyclohexane 99,5 GC_ASSAY_METHOD, EMSURE® Reag. Ph. Eur., ACS, ISO analysis_for, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Hexahydrobenzene, Hexamethylene

1.09666.1000 1.09666.2500 1.09666.6010 1.09666.6025
CA1.09666.1000CS 1173 CAD
CA1.09666.1000 CA1.09666.2500 CA1.09666.6010 CA1.09666.6025
Cyclohexane 99,5 GC_ASSAY_METHOD, EMSURE® Reag. Ph. Eur., ACS, ISO analysis_for, Supelco®
Cyclohexane
Formula: C₆H₁₂
MW: 84.16 g/mol
Boiling Pt: 81 (lit.)
Melting Pt: 7 (lit.)
Density: 0.779 g/ml (25°C) (lit.)
Flash Pt: –18 °C
Storage Temperature: Ambient
MDL Number: MFCD00003814
CAS Number: 110-82-7
EINECS: 203-806-2
UN: 1145
ADR: 3 II
Merck Index: 13,02752
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a1e51ba998bef126804c3fd25b361be72cd9545&vid=d1e7f75d9f2a944eb15a7740c5254588cc9a4e67&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 55 mg/l (20 °C)
Melting Point 6 °C
Molar Mass 84.16 g/mol
Boiling Point 81 °C (1013 hPa)
Vapor Pressure 103 hPa (20 °C)
Flash Point -18 °C
Refractive Index 1.4264 (20 °C)
Explosion Limit 1.2 - 8.3 %(V)
Density 0.78 g/cm3 (20 °C)
pH (H2O) not applicable
Ignition Point 260 °C
Viscosity kinematic 1.26 mm2/s (20 °C)
Evaporation number 3.5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR