Butyl acetate ≥99.5% (by GC), EMSURE® for analysis, Supelco®
Supplier: MilliporeSigma
|
Warning
|
1.09652.1000
1.09652.2500
CA1.09652.1000CS
1288
CAD
CA1.09652.1000
CA1.09652.2500
Butyl acetate ≥99.5% (by GC), EMSURE® for analysis, Supelco®
Butyl acetate
Formula:
CH₃COO(CH₂)₃CH₃ Boiling Pt: 126 °C (1013 hPa) Melting Pt: –76 °C Density: 0.875 g/cm³ (20 °C) Flash Pt: 25 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009445 CAS Number: 123-86-4 EINECS: 204-658-1 UN: 1123 ADR: 3, Merck Index: 13,01534 |
|
Specification Test Results
Purity (GC) | ≥ 99.5 % |
Identity (IR) | conforms |
Color | ≤ 10 Hazen |
Acidity | ≤ 0.0017 meq/g |
Readily carbonizable substances | conforms |
n-Butyl alcohol (GC) | ≤ 0.2 % |
n-Butyl formiate (GC) | ≤ 0.1 % |
n-Butyl propionate (GC) | ≤ 0.1 % |
Evaporation residue | ≤ 0.001 % |
Water | ≤ 0.1 % |
Al (Aluminium) | ≤ 0.00005 % |
B (Boron) | ≤ 0.000002 % |
Ba (Barium) | ≤ 0.00001 % |
Ca (Calcium) | ≤ 0.00005 % |
Cd (Cadmium) | ≤ 0.000005 % |
Co (Cobalt) | ≤ 0.000002 % |
Cr (Chromium) | ≤ 0.000002 % |
Cu (Copper) | ≤ 0.000002 % |
Fe (Iron) | ≤ 0.00001 % |
Mg (Magnesium) | ≤ 0.00001 % |
Mn (Manganese) | ≤ 0.000002 % |
Ni (Nickel) | ≤ 0.000002 % |
Pb (Lead) | ≤ 0.00001 % |
Sn (Tin) | ≤ 0.00001 % |
Zn (Zinc) | ≤ 0.00001 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...