2-Propanol ≥99.8% (by GC), EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Supplier: MilliporeSigma
|
Danger
|
1.09634.1011
1.09634.2500
1.09634.2511
1.09634.5000
1.09634.6010
1.09634.6025
1.09634.6190
1.09634.9180
1.09634.1000
CA1.09634.1011CS
696
CAD
CA1.09634.1011
CA1.09634.2500
CA1.09634.2511
CA1.09634.5000
CA1.09634.6010
CA1.09634.6025
CA1.09634.6190
CA1.09634.9180
CA1.09634.1000
2-Propanol ≥99.8% (by GC), EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
2-Propanol
Formula:
(CH₃)₂CHOH Boiling Pt: 82 °C (lit.) Melting Pt: –89.5 °C (lit.) Density: 0.785 g/mL (25 °C) (lit.) Flash Pt: 12 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011674 CAS Number: 67-63-0 EINECS: 200-661-7 UN: 1219 ADR: 3,II REACH: 01-2119457558-25 Merck Index: 13,05228 |
|
Specification Test Results
Purity (GC) | ≥ 99.8 % |
Identity (IR) | conforms |
Appearance | clear |
Color | ≤ 10 Hazen |
Solubility in water | conforms |
Acidity | ≤ 0.0001 meq/g |
Alkalinity | ≤ 0.0001 meq/g |
Density (d 20 °C/20 °C) | 0.784 - 0.787 |
Boiling point | 81 - 83 °C |
Chloride (Cl) | ≤ 300 ppb |
Nitrate (NO₃) | ≤ 300 ppb |
Phosphate (PO₄) | ≤ 500 ppb |
Sulfate (SO₄) | ≤ 1000 ppb |
Substances reducing potassium permanganate (as O) | ≤ 2 ppm |
Carbonyl compounds (as CO) | ≤ 0.002 % |
Matter discolored by H₂SO₄ | ≤ 10 Hazen |
Acetone (GC) | ≤ 0.01 % |
Ethanol (GC) | ≤ 0.01 % |
Isopropylether (GC) | ≤ 0.01 % |
Methanol (GC) | ≤ 0.01 % |
1-Propylalcohol (GC) | ≤ 0.1 % |
Ag (Silver) | ≤ 0.000002 % |
Al (Aluminium) | ≤ 0.00005 % |
As (Arsenic) | ≤ 0.000002 % |
Au (Gold) | ≤ 0.000002 % |
B (Boron) | ≤ 0.000002 % |
Ba (Barium) | ≤ 0.00001 % |
Be (Beryllium) | ≤ 0.000002 % |
Bi (Bismuth) | ≤ 0.00001 % |
Ca (Calcium) | ≤ 0.00005 % |
Cd (Cadmium) | ≤ 0.000005 % |
Co (Cobalt) | ≤ 0.000002 % |
Cr (Chromium) | ≤ 0.000002 % |
Cu (Copper) | ≤ 0.000002 % |
Fe (Iron) | ≤ 0.00001 % |
Ga (Gallium) | ≤ 0.000002 % |
In (Indium) | ≤ 0.000002 % |
Li (Lithium) | ≤ 0.000005 % |
Mg (Magnesium) | ≤ 0.00001 % |
Mn (Manganese) | ≤ 0.000002 % |
Mo (Molybdenum) | ≤ 0.000002 % |
Ni (Nickel) | ≤ 0.000002 % |
Pb (Lead) | ≤ 0.00001 % |
Pt (Platinum) | ≤ 0.000002 % |
Sb (Antimony) | ≤ 0.000002 % |
Sn (Tin) | ≤ 0.00001 % |
Ti (Titanium) | ≤ 0.000002 % |
Tl (Thallium) | ≤ 0.000002 % |
V (Vanadium) | ≤ 0.000002 % |
Zn (Zinc) | ≤ 0.00001 % |
Zr (Zirconium) | ≤ 0.000002 % |
Evaporation residue | ≤ 0.001 % |
Water | ≤ 0.05 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...