(±)-2-Butanol, EMSURE® for analysis, Supelco®
Supplier: MilliporeSigma
|
Warning
|
1.09630.1000
1.09630.2500
CA1.09630.1000CS
901
CAD
CA1.09630.1000
CA1.09630.2500
(±)-2-Butanol, EMSURE® for analysis, Supelco®
(±)-2-Butanol
Formula:
H₃CCH₂CH(OH)CH₃ Boiling Pt: 94 °C (1013 hPa) Melting Pt: –115 °C Density: 0.8534 g/cm³ (20 °C) Flash Pt: 23 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004569 CAS Number: 78-92-2 EINECS: 201-158-5 UN: 1120 ADR: 3, Merck Index: 13,01540 |
|
Specification Test Results
Solubility | 125 g/l (20 °C) |
Melting Point | -115 °C |
Molar Mass | 74.12 g/mol |
Boiling Point | 99 - 102 °C (1013 hPa) |
Vapor Pressure | 16.5 hPa (20 °C) |
Flash Point | 24 °C |
Explosion Limit | 1.4 - 9.8 %(V) |
Density | 0.81 g/cm3 (20 °C) |
pH | (H2O, 20 °C) neutral |
Ignition Point | 390 °C |
Evaporation number | 20 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...